Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Carboxypeptidase A1/CPA1, Mouse anti-Human, Clone: CPA1/2712, Novus Biologicals™
Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP27974520UG
Description
Carboxypeptidase A1/CPA1 Monoclonal specifically detects Carboxypeptidase A1/CPA1 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin, Protein Array.Specifications
Carboxypeptidase A1/CPA1 | |
Monoclonal | |
0.2 mg/mL | |
Immunohistochemistry 1-2 ug/ml, Immunohistochemistry-Paraffin 1-2 ug/ml, Protein Array | |
P15085, P15085 | |
CPA1 | |
Recombinant full-length human Carboxypeptidase A1/CPA1 protein | |
20 μg | |
Mast Cell Markers | |
1357 | |
Store at 4°C. | |
IgG2c κ |
Immunohistochemistry, Immunohistochemistry (Paraffin), Peptide Array | |
CPA1/2712 | |
Unconjugated | |
10 mM PBS with 0.05% BSA | |
carboxypeptidase A1, carboxypeptidase A1 (pancreatic), CPA, EC 3.4.17, EC 3.4.17.1 | |
Mouse | |
Protein A or G purified | |
RUO | |
Primary | |
Human | |
Purified |
Provide Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction