Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Carboxypeptidase A1/CPA1 Antibody (OTI2A3), Janelia Fluor™ 646, Novus Biologicals™
Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP270431JF646
Description
Carboxypeptidase A1/CPA1 Monoclonal specifically detects Carboxypeptidase A1/CPA1 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence.Specifications
Carboxypeptidase A1/CPA1 | |
Monoclonal | |
Janelia Fluor 646 | |
50mM Sodium Borate with 0.05% Sodium Azide | |
CPA1 | |
Recombinant protein expressed in E.coli corresponding to amino acids 1-419 of human Carboxypeptidase A1/CPA1. | |
0.1 mL | |
Primary | |
Human | |
IgG2a |
Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence | |
OTI2A3 | |
Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence | |
carboxypeptidase A1, carboxypeptidase A1 (pancreatic), CPA, EC 3.4.17, EC 3.4.17.1 | |
Mouse | |
Affinity Purified | |
Mast Cell Markers | |
1357.0 | |
Store at 4°C in the dark. |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction