Learn More
Carboxypeptidase A2/CPA2 Antibody (OTI1C7), DyLight 755, Novus Biologicals™
SDP
Mouse Monoclonal Antibody
Supplier:  Novus Biologicals NBP270440IR
Catalog No. NBP270440IR
Add to Cart
Description
Description
Carboxypeptidase A2/CPA2 Monoclonal specifically detects Carboxypeptidase A2/CPA2 in Human, Mouse, Rat samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence.
Specifications
Specifications
Carboxypeptidase A2/CPA2
Monoclonal
DyLight 755
50mM Sodium Borate with 0.05% Sodium Azide
CPA2
Full length recombinant protein of human Carboxypeptidase A2/CPA2 (NP_001860) produced in HEK293T cells.
0.1 mL
1358.0
Store at 4°C in the dark.
Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry, Immunofluorescence
OTI1C7
Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence
carboxypeptidase A2, carboxypeptidase A2 (pancreatic), EC 3.4.17, EC 3.4.17.15
Mouse
Affinity Purified
Primary
Human, Mouse, Rat
IgG2b
Product Suggestions
Product Suggestions