Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Novus Biologicals™ Carboxypeptidase A2/CPA2 Antibody Pair [HRP]

Monoclonal Antibody Pair

Manufacturer:  Novus Biologicals™ NBP2794755PLATES

 View more versions of this product

Catalog No. NB009990


Add to cart

Description

Description

This solid phase sandwich ELISA utilizes a monoclonal antibody specific for Human Carboxypeptidase A2/CPA2.
Specifications

Specifications

Sandwich ELISA
Antibody Pairs
12.5 pg/mL
1358
RUO
Carboxypeptidase A2/CPA2
5 Plates
12.5-800 pg/mL
carboxypeptidase A2, carboxypeptidase A2 (pancreatic), EC 3.4.17, EC 3.4.17.15
HRP
This solid phase sandwich ELISA utilizes a monoclonal antibody specific for Human Carboxypeptidase A2/CPA2.
Sandwich ELISA
  • Detection Antibody: 0.5 mg/mL rabbit anti-Carboxypeptidase A2/CPA2 monoclonal antibody conjugated to horseradish-peroxidase (HRP).
  • Capture Antibody: 1.0 mg/mL of mouse anti-Carboxypeptidase A2/CPA2 monoclonal antibody,
  • Standard: Each vial contains 50 ng of recombinant Carboxypeptidase A2/CPA2.
  • Storage is content dependent.
    Rabbit/Mouse
    Human
    CPA2
    1. Capture Antibody: Dilute to a working concentration of 2 μg/mL in CBS before coating. 2. Detection Antibody: Dilute to working concentration of 0.25 μg/mL in detection antibody dilution buffer before use. 3. Standard: Reconstitute with 1 ml detection antibody dilution buffer. After reconstitution, store at -20 to -80°C in a manual defrost freezer. A seven-point standard curve usi ng 2-fold serial dilutions in sample dilution buffer, and a high standard of 800 pg/mL is recommended.
    SDS
    Documents
    Product Certifications
    Promotions

    Promotions

    Provide Content Correction

    We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

    Product Title

    By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

    Cancel Submit