Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Carboxypeptidase A2/CPA2 Mouse anti-Human, Mouse, Rat, Janelia Fluor™ 549, Clone: OTI3D8, Novus Biologicals™

Mouse Monoclonal Antibody

Manufacturer:  Novus Biologicals NBP270443JF549

Catalog No. N270443J549


Add to cart

Description

Description

Carboxypeptidase A2/CPA2 Monoclonal antibody specifically detects Carboxypeptidase A2/CPA2 in Human, Mouse, Rat samples. It is validated for Immunohistochemistry.
Specifications

Specifications

Carboxypeptidase A2/CPA2
OTI3D8
Immunohistochemistry
Affinity Purified, Conjugated
CPA2
Full length recombinant protein of human Carboxypeptidase A2/CPA2 (NP_001860) produced in HEK293T cells.
Immunogen affinity purified
Store at 4°C in the dark.
Monoclonal
Human, Mouse, Rat
Immunohistochemistry
Janelia Fluor 549
50mM Sodium Borate with 0.05% Sodium Azide
carboxypeptidase A2, carboxypeptidase A2 (pancreatic), EC 3.4.17, EC 3.4.17.15
Mouse
IgG1
0.1 mL
Primary
1358.0
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit