Learn More
Carboxypeptidase A2 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech Non-distribution product as customer accommodation.
Rabbit Polyclonal Antibody
Manufacturer:  Proteintech Group IncSupplier Diversity Partner 156261AP
Catalog No. 50-173-0362
Add to cart
Description
Description
Three different forms of human pancreatic procarboxypeptidase A have been isolated. The encoded protein represents the A2 form, which is a monomeric protein with different biochemical properties from the A1 and A3 forms. The A2 form of pancreatic procarboxypeptidase acts on aromatic C-terminal residues and is a secreted protein.
Specifications
Specifications
Carboxypeptidase A2
0.18 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
P19222, P48052, Q504N0
Carboxypeptidase A2, CPA2
Rabbit
IgG
150 μL
-20°C
Primary
1358, 232680, 296959
Immunohistochemistry (Paraffin), Western Blot
Unconjugated
CPA2
Liquid
CPA2
Carboxypeptidase A2 Fusion Protein Ag8080
Antigen Affinity Chromatography
RUO
Antibody
Polyclonal
Human, Mouse, Rat