Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Novus Biologicals Carboxypeptidase B1/CPB1 Antibody Pair [HRP]
Rabbit Monoclonal Antibody Pair
Manufacturer: Novus Biologicals NBP2795705PLATES
Description
This solid phase sandwich ELISA utilizes a monoclonal antibody specific for Mouse Carboxypeptidase B1/CPB1.Specifications
12.5 pg/mL | |
Conjugated | |
1360 | |
Antibody Pairs | |
12.5-800 pg/mL | |
Storage is content dependent. | |
Rabbit | |
Mouse | |
CPB1 | |
1. Capture Antibody: Dilute to a working concentration of 2 μg/mL in CBS before coating. 2. Detection Antibody: Dilute to working concentration of 0.5 μg/mL in detection antibody dilution buffer before use. 3. Standard: Reconstitute with 1 ml detection antibody dilution buffer. After reconstitution, store at -20 to -80°C in a manual defrost freezer. A seven-point standard curve using 2-fold serial dilutions in sample dilution buffer, and a high standard of 0.8 ng/mL is recommended. |
HRP | |
Sandwich ELISA | |
RUO | |
This solid phase sandwich ELISA utilizes a monoclonal antibody specific for Mouse Carboxypeptidase B1/CPB1. | |
Carboxypeptidase B1/CPB1 | |
5 Plates | |
Sandwich ELISA | |
carboxypeptidase B, carboxypeptidase B1 (tissue), CPB, DKFZp779K1333, EC 3.4.17, EC 3.4.17.2, Pancreas-specific protein, PASP, PCPB, procarboxypeptidase B |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title