Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Novus Biologicals™ Carboxypeptidase B1/CPB1 Antibody Pair [HRP]

Monoclonal Antibody Pair

Manufacturer:  Novus Biologicals™ NBP2794765PLATES

 View more versions of this product

Catalog No. NB009992


Add to cart

Description

Description

This solid phase sandwich ELISA utilizes a monoclonal antibody specific for Human Carboxypeptidase B1/CPB1.
Specifications

Specifications

HRP
Antibody Pairs
6.25 pg/mL
1360
RUO
Carboxypeptidase B1/CPB1
5 Plates
6.25-400 pg/mL
carboxypeptidase B, carboxypeptidase B1 (tissue), CPB, DKFZp779K1333, EC 3.4.17, EC 3.4.17.2, Pancreas-specific protein, PASP, PCPB, procarboxypeptidase B
Sandwich ELISA
This solid phase sandwich ELISA utilizes a monoclonal antibody specific for Human Carboxypeptidase B1/CPB1.
Sandwich ELISA
  • Standard: Each vial contains 30 ng of recombinant Carboxypeptidase B1/CPB1.
  • Capture Antibody: 0.2 mg/mL of mouse anti-Carboxypeptidase B1/CPB1 monoclonal antibody,
  • Detection Antibody: 0.5 mg/mL of rabbit anti-Carboxypeptidase B1/CPB1 monoclonal antibody conjugated to horseradish-peroxidase (HRP).
  • Storage is content dependent.
    Rabbit/Mouse
    Human
    CPB1
    1. Capture Antibody: Dilute to a working concentration of 2 μg/mL in CBS before coating. 2. Detection Antibody: Dilute to working concentration of 0.25 μg/mL in detection antibody dilution buffer before use. 3. Standard: Reconstitute with 1 ml detection antibody dilution buffer. After reconstitution, store at -20 to -80°C in a manual defrost freezer. A seven-point standard curve usi ng 2-fold serial dilutions in sample dilution buffer, and a high standard of 400 pg/mL is recommended.
    SDS
    Documents
    Product Certifications
    Promotions

    Promotions

    Provide Content Correction

    We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

    Product Title

    By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

    Cancel Submit