Learn More
Carboxypeptidase B2/CPB2 Mouse anti-Human, Alexa Fluor 647, Clone: 13D5, Novus Biologicals™
Mouse Monoclonal Antibody
Manufacturer:  Novus Biologicals NBP179045AF647
Catalog No. NBP179045AH
Add to cart
Description
Description
Carboxypeptidase B2/CPB2 Monoclonal antibody specifically detects Antigen in Human samples. It is validated for Enzyme Linked Immunosorbent Assay.
Specifications
Specifications
Carboxypeptidase B2/CPB2
13D5
ELISA
Purified
CPB2
Human TAF
Protein A purified
RUO
Primary
1361
Human
ELISA
Alexa Fluor 647
50mM Sodium Borate with 0.05% Sodium Azide
carboxypeptidase B2, carboxypeptidase B2 (plasma), carboxypeptidase B2 (plasma, carboxypeptidase U), Carboxypeptidase U, CPUEC 3.4.17.20, EC 3.4.17, PCPB, Plasma carboxypeptidase B, TAFIcarboxypeptidase B-like protein, Thrombin-activable fibrinolysis inhibitor, thrombin-activatable fibrinolysis inhibitor
Mouse
IgG1
0.2 ml
Store at 4C in the dark.
Monoclonal
Human TAFI