Learn More
Carboxypeptidase B2/CPB2 Mouse anti-Human, HRP, Clone: 13D5, Novus Biologicals™
Mouse Monoclonal Antibody
Manufacturer:  Novus Biologicals NBP179045H
Catalog No. NBP179045H
Add to cart
Description
Description
Carboxypeptidase B2/CPB2 Monoclonal antibody specifically detects Antigen in Human samples. It is validated for Enzyme Linked Immunosorbent Assay.
Specifications
Specifications
Carboxypeptidase B2/CPB2
13D5
ELISA
Purified
CPB2
Human TAF
Protein A purified
RUO
Primary
1361
Human
ELISA
HRP
PBS with No Preservative
carboxypeptidase B2, carboxypeptidase B2 (plasma), carboxypeptidase B2 (plasma, carboxypeptidase U), Carboxypeptidase U, CPUEC 3.4.17.20, EC 3.4.17, PCPB, Plasma carboxypeptidase B, TAFIcarboxypeptidase B-like protein, Thrombin-activable fibrinolysis inhibitor, thrombin-activatable fibrinolysis inhibitor
Mouse
IgG1
0.2 ml
Store at 4C in the dark.
Monoclonal
Human TAFI