Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Carboxypeptidase M Antibody (OTI2D11), DyLight 550, Novus Biologicals™
SDP

Mouse Monoclonal Antibody

Supplier:  Novus Biologicals NBP270446R

Catalog No. NBP270446R


Add to Cart

Description

Description

Carboxypeptidase M Monoclonal specifically detects Carboxypeptidase M in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry.
Specifications

Specifications

Carboxypeptidase M
Monoclonal
DyLight 550
50mM Sodium Borate with 0.05% Sodium Azide
CPM
Recombinant protein fragment corresponding to amino acids 124-423 of human Carboxypeptidase M (NP_001865) produced in E.coli.
0.1 mL
1368.0
Store at 4°C in the dark.
Western Blot, Immunohistochemistry
OTI2D11
Western Blot, Immunohistochemistry
carboxypeptidase M, EC 3.4.17, EC 3.4.17.12
Mouse
Affinity Purified
Primary
Human, Mouse, Rat
IgG2b
Product Suggestions

Product Suggestions

SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit