Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Carboxypeptidase M Antibody (OTI2D11), Alexa Fluor™ 405, Novus Biologicals™
Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP270446AF405
Description
Carboxypeptidase M Monoclonal specifically detects Carboxypeptidase M in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry.Specifications
Carboxypeptidase M | |
Monoclonal | |
Alexa Fluor 405 | |
50mM Sodium Borate with 0.05% Sodium Azide | |
CPM | |
Recombinant protein fragment corresponding to amino acids 124-423 of human Carboxypeptidase M (NP_001865) produced in E.coli. | |
0.1 mL | |
1368.0 | |
Store at 4°C in the dark. |
Western Blot, Immunohistochemistry | |
OTI2D11 | |
Western Blot, Immunohistochemistry | |
carboxypeptidase M, EC 3.4.17, EC 3.4.17.12 | |
Mouse | |
Affinity Purified | |
Primary | |
Human, Mouse, Rat | |
IgG2b |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction