Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Carboxypeptidase M Mouse anti-Human, Janelia Fluor™ 646, Clone: OTI2D11, Novus Biologicals™

Mouse Monoclonal Antibody

Manufacturer:  Novus Biologicals NBP270446JF646

Catalog No. N270446J646


Add to cart

Description

Description

Carboxypeptidase M Monoclonal antibody specifically detects Carboxypeptidase M in Human samples. It is validated for Immunohistochemistry.
Specifications

Specifications

Carboxypeptidase M
OTI2D11
Immunohistochemistry
Affinity Purified, Conjugated
CPM
Recombinant protein fragment corresponding to amino acids 124-423 of human Carboxypeptidase M (NP_001865) produced in E.coli.
Immunogen affinity purified
Store at 4°C in the dark.
Monoclonal
Human
Immunohistochemistry
Janelia Fluor 646
50mM Sodium Borate with 0.05% Sodium Azide
carboxypeptidase M, EC 3.4.17, EC 3.4.17.12
Mouse
IgG2b
0.1 mL
Primary
1368.0
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit