Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Carboxypeptidase Y, Yeast, Affymetrix/USB™

Click to view available options
Quantity:
1mg
10mg
Packaging:
Plastic bottle
9046-67-7
MFCD00130723
E.C.3.4.16.5
9046-67-7
MFCD00130723
E.C.3.4.16.5

Useful enzyme in sequence analysis of proteins and peptides

Quantity 10mg
Packaging Plastic bottle
Physical Form Lyophilized Powder
Chemical Name or Material Carboxypeptidase Y