Learn More
Thermo Scientific Lab Vision CEA (Carcinoembryonic Antigen)/CD66e Ab-3, Mouse Monoclonal Antibody
Ensure accurate, reproducible results in immunohistochemistry, western blotting, immunofluorescence and immunoprecipitation experiments.
Manufacturer: Thermo Scientific MS613R1
Description
Ensure accurate, reproducible results in immunohistochemistry, western blotting, immunofluorescence and immunoprecipitation experiments with Thermo Scientific CEA (Carcinoembryonic Antigen)/CD66e Ab-3, Mouse Monoclonal Antibody.
Host Species: Mouse
Clone: COL-1
Isotype: IgG2a/κ
Species Reactivity: Human. Does not react with rat. Others not known.
Immunogen: Human colon carcinoma extract
Molecular Weight: 180 kDa
Positive Control: LS174T cells, Colon CA
Cellular Localization: Cytoplasmic and luminal membrane
Recommended for:
- Immunohistochemistry (Formalin/paraffin)
- Western Blotting
- Immunofluorescence
- Immunoprecipitation (Native and denatured)
Note:
USA: RUO; Int'l: RUO
Specifications
CEA (Carcinoembryonic Antigen)/CD66e Ab-3 | |
Monoclonal | |
Unconjugated | |
Prediluted | |
IgG2a, κ | |
RUO | |
Primary | |
Monoclonal |
Immunofluorescence, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot | |
COL-1 | |
Ready-to-use for IHC (UVLP) | |
Murine | |
1mL | |
Oncogenesis | |
Primary | |
Human |
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.