Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Carcinoma Glycans Mouse anti-Human, Unconjugated, Clone: mAb-A4, MilliporeSigma™

Mouse Monoclonal Antibody

$153.50 - $386.00

Specifications

Antigen Carcinoma Glycans
Applications Flow Cytometry, Immunohistochemistry, Immunoprecipitation, Western Blot
Conjugate Unconjugated
Format Purified
Host Species Mouse
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
MABC17091MI
View Documents
MilliporeSigma
MABC1709100UG
100 μg
Each for $386.00
Add to cart
 
MABC17092MI
View Documents
MilliporeSigma
MABC170925UG
25 μg
Each for $153.50
Add to cart
 
Description

Description

Carcinoma Glycans Monoclonal antibody specifically detects Carcinoma Glycans in Human samples. It is validated for Flow Cytometry, Immunohistochemistry, Immunoprecipitation, Western Blotting.
Specifications

Specifications

Carcinoma Glycans
Unconjugated
Mouse
Cancer Cell Glycans
Monoclonal
Protein G Purified
Flow Cytometry, Immunohistochemistry, Immunoprecipitation, Western Blot
Purified
Human
Live HES-3 human embryonic stem cells.
Primary
Stable for 1 year at 2 to 8°C from date of receipt.
SDS
Product Certifications
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit