Learn More
Enzo Life SciencesSupplier Diversity Partner CARD 9 polyclonal antibody (100 µg)  DFS Item
Manufacturer:  Enzo Life SciencesSupplier Diversity Partner ADI905188100
Host: Rabbit. Immunogen: Synthetic peptide corresponding to aa 521-536 of human CARD-9. UniProt: Q9H257. Species Reactivity: Human. Applications: ICC, WB. Recommended dilutions: Immunocytochemistry (10µg/ml). Western Blot (0.5-1.0µg/ml). Suggested dilutions/conditions may not be available for all applications. Optimal conditions must be determined individually for each application. Purity: Peptide affinity purified. Formulation: Liquid. In PBS containing 0.02% sodium azide.
Catalog No. 50-658-283