Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CARD11/CARMA1 Antibody (SR1136), Novus Biologicals™

Rabbit Monoclonal Antibody
$309.50 - $423.00
Specifications
Antigen | CARD11/CARMA1 |
---|---|
Clone | SR1136 |
Dilution | Western Blot 1:500-1:2000, Immunocytochemistry/Immunofluorescence 1:50-1:200 |
Applications | Western Blot, Immunofluorescence |
Classification | Monoclonal |
Description
CARD11/CARMA1 Monoclonal antibody specifically detects CARD11/CARMA1 in Human, Mouse samples. It is validated for Western Blot, ImmunofluorescenceSpecifications
CARD11/CARMA1 | |
Western Blot 1:500-1:2000, Immunocytochemistry/Immunofluorescence 1:50-1:200 | |
Monoclonal | |
Purified | |
RUO | |
Human, Mouse | |
BIMP3card-maguk protein 1, CARD-containing MAGUK protein 1, Carma 1, CARMA1bcl10-interacting maguk protein 3, caspase recruitment domain family, member 11, caspase recruitment domain-containing protein 11, MGC133069 | |
A synthesized peptide derived from human CARD11/CARMA1 (Uniprot #: Q9BXL7) | |
Primary | |
Store at -20° C. Avoid freeze/thaw cycles. |
SR1136 | |
Western Blot, Immunofluorescence | |
Unconjugated | |
Rabbit | |
Apoptosis | |
PBS, pH 7.4, 150mM NaCl, 50% glycerol. | |
84433 | |
IgG | |
Affinity purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title