Learn More
CARD11 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech Non-distribution product as customer accommodation.
Rabbit Polyclonal Antibody
Manufacturer:  Proteintech Group IncSupplier Diversity Partner 217411AP
Catalog No. 50-173-3573
Add to cart
Description
Description
The protein encoded by this gene belongs to the membrane-associated guanylate kinase (MAGUK) family, a class of proteins that functions as molecular scaffolds for the assembly of multiprotein complexes at specialized regions of the plasma membrane. This protein is also a member of the CARD protein family, which is defined by carrying a characteristic caspase-associated recruitment domain (CARD). This protein has a domain structure similar to that of CARD14 protein. The CARD domains of both proteins have been shown to specifically interact with BCL10, a protein known to function as a positive regulator of cell apoptosis and NF-kappaB activation. When expressed in cells, this protein activated NF-kappaB and induced the phosphorylation of BCL10.
Specifications
Specifications
CARD11
0.26 mg/mL
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
Q8CIS0, Q9BXL7
BIMP3, CARD11, Carma 1, CARMA1
Rabbit
IgG
150 μL
-20°C
Primary
100363332, 108723, 84433
Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
Unconjugated
CARD11
Liquid
Card11
CARD11 Fusion Protein Ag16468
Antigen Affinity Chromatography
RUO
Antibody
Polyclonal
Human, Mouse, Rat