Learn More
CARD12 Rabbit anti-Human, Mouse, DyLight™ 350, Polyclonal, Novus Biologicals™
Rabbit Polyclonal Antibody
Manufacturer:  Novus Biologicals NBP178980UV
Catalog No. NBP178980UV
Add to cart
Description
Description
CARD12 Polyclonal antibody specifically detects CARD12 in Human,Mouse samples. It is validated for Immunohistochemistry,Immunocytochemistry,Immunofluorescence,Immunohistochemistry (Paraffin).
Specifications
Specifications
CARD12
DyLight™ 350
50 mM sodium borate with 0.05% sodium azide
NLRC4
A synthetic peptide made to a C-terminal portion of the human CARD12 protein (between residues 950-1015) [Uniprot: Q9NPP4]
0.1 mL
Primary
58484
Immunocytochemistry, Immunofluorescence, Immunohistochemistry, Immunohistochemistry (Paraffin)
Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin
CARD12leucine rich repeat and CARD domaincontaining 4, caspase recruitment domain family, member 12, Caspase recruitment domain-containing protein 12, Clan protein, CLAN1ipaf, CLANCARD, LRR, and NACHT-containing protein, CLR2.1, Ice protease-activating factor, Ipaf, NLR family, CARD domain containing 4
Rabbit
IgG
Store at 4°C in the dark.
Polyclonal
Human, Mouse