Learn More
CARD6 Rabbit anti-Human, Polyclonal, Invitrogen™
Rabbit Polyclonal Antibody
Manufacturer:  Invitrogen PA549737
Catalog No. PA549737
Add to cart
Description
Description
The antibody detects endogenous levels of total CARD6 protein.
This gene encodes a protein that contains a caspase recruitment domain (CARD), an antiparallel six-helical bundle that mediates homotypic protein-protein interactions. The encoded protein is a microtubule-associated protein that has been shown to interact with receptor-interacting protein kinases and positively modulate signal transduction pathways converging on activation of the inducible transcription factor NF-kB.
Specifications
Specifications
CARD6
Polyclonal
Unconjugated
CARD6
Liquid
CARD6
Synthesized peptide derived from human CARD6.
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human
Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
1 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Q9BX69
Caspase recruitment domain-containing protein 6, CARD6,
Rabbit
IgG
100 μL
-20°C
Primary
84674