Learn More
Cardiac Science™ AED Trainer
More instructive and intuitive voice prompts, improved voice quality and clarity in a user-friendly form factor
Manufacturer:  Cardiac Science™ 1805020001
Catalog No. 19-171-013
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Part Not Found
Includes: Each AED trainer supplied with one set of reusable training electrode pads; an infrared, wireless remote; a control panel that simulates fully automataic models; and a user manual.
This item is not returnable. View return policy
Description
Description
Wireless infrared remote control for teaching both semi-automatic and fully automatic defibrillation. Versatile design works with Powerheart™ AED G3, Powerheart AED G3 Automatic, and FirstSave™ AED G3
  • Four preprogrammed, simulated rescue scenarios which support American Heart Association (AHA) guidelines and AHA Heartsaver AED curriculum
  • Rescue scenarios can be changed and simulated cardiac rhythms varied from shockable to nonshockable
  • Instructor advances rescue stages through an infrared, wireless remote control and may pause at any time for additional instruction
  • Students hear voice prompts that would typically occur during an actual rescue attempt
  • Preprogrammed with multiple languages
  • Economical to use—battery powered (see below) and compatible with any type of CPR manikin
  • L x W x H: 1.1 x 8.3 x 2.8 in. (29 x 21 x 7cm)
Order Info
Requires two each “D” and “AAA” batteries, not included.
Compliance
Optimized with AHA guidelines
Specifications
Specifications
AED G3 Trainer
7cm
Teaching both semi-automatic and fully automatic defibrillation, versatile design works with Powerheart™ AED G3, Powerheart AED G3 automatic and FirstSave™ AED G3
2.8 in.
Each AED trainer supplied with one set of reusable training electrode pads an infrared, wireless remote, a control panel that simulates fully automatic models and a user manual