Learn More
Cardiac Science™ Pediatric Defibrillation Electrodes for Powerheart AED G3 defibrillators
Can be used to monitor patient for up to two hours and to deliver up to 15 shocks
Manufacturer:  Cardiac Science™ 9730002
Catalog No. 19-120-2468
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Part Not Found
This item is not returnable. View return policy
Description
Description
  • Recommended for children under 8 years old or weighing less than 25kg (55 lb.)
  • Reduce defibrillator energy output to prescribed 50 joule level
  • Two-year shelf life
Pads cannot be pre-connected to AED and must be connected immediately before use.
Specifications
Specifications
Used to monitor patient for up to two hours and to deliver up to 15 shocks
6 lb.
Pediatric defibrillation electrodes
Powerheart™ AED G3 defibrillators
2.7kg
Safety and Handling
Safety and Handling
ShelfLife : 2 Year(s)