Learn More
Cardiac Science™ Powerheart™ AED G3 Plus
Extensive prompts coach user through AED use
Manufacturer:  Cardiac Science™ 9390A1001WP
 View more versions of this product
Catalog No. 19-172-008
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Part Not Found
Includes: Defibrillator, Intellisense™ battery; pair of defibrillation pads; Quick Start Tool Kit which includes CD-Rom with AED Manual, Training Video, RescueLink™ and MDLink™; and serial communication cable.
This item is not returnable. View return policy
Description
Description
Reliable, easy-to-use Powerheart AED G3 Plus automates the delivery of life-saving therapy for sudden cardiac arrest with a unique combination of features
  • Features Rescue Coach™ intuitive voice prompts that guide rescuers step-by-step through the rescue process while following AHA/ERC 2005 Guidelines
  • Patented RescueReady™ technology self-tests battery, system electronics, and electrodes (presence and functionality) on a daily, weekly, and monthly basis
  • Integrated metronome for proper CPR compression and rescue breath pacing
  • Easy-to-read display for use in noisy environments or by rescuers who are hearing impaired
  • Patented non-polarized interchangeable defibrillation pads simplify pad placement
  • Offers configurable STAR™ (Self-Tracking Active Response) biphasic energy protocols
  • Fully automatic (no-button operation) and semi-automatic (one-button operation) models
Order Info
Accessories available. (See Cat. No. 19—172–011)
Warranty and Services
  • Seven-year AED warranty
  • Four-year full replacement battery warranty
Specifications
Specifications
External defibrillator (AED), Wall package
95J to 351J
3.3 in.
(1) 9390A Powerheart AED G3 Plus Full-Auto; (1) 9146 IntelliSense Battery; (2) pairs 9131 adult defibrillation pads; (1) carry bag; (1) 5550 ready kit; (1) Quick Start Tool Kit (includes Quick Start Guide, CD-ROM with AED manual, training video, RescueLink & MDLink); (1) serial communications cable; (1) 50-00392-20 wall cabinet wwith alarm
Wall mount
3.1kg
27cm
6.6 lb.
For lay rescuers to use in public access defibrillation areas such as schools, athletic fields, and workplace environments.
8cm
60 minutes ECG data with event annotation, multiple rescue functionality
Fully automatic
10.6 in.