Learn More
Cardiac Science Powerheart AED G3 Pro
Rescue Ready performance provides healthcare and rescue professionals extended capabilities for emergency treatment of sudden cardiac arrest followed by continuous cardiac monitoring
Specifications
Product Type | Automated External Defibrillator |
---|
Includes: Defibrillator, one pair of defibrillation pads, and Quick Start Tool Kit ( includes Quick Start Guide, CD-ROM with AED Manual, Training Video, Rescuelink and MDLink software). 19-167-229 includes a nonrechargeable IntelliSense lithium battery that provides up to 12 hours of use. 19-230-1130 includes a rechargeable battery.
Catalog Number | Mfr. No. | Includes | Weight (Metric) | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Includes | Weight (Metric) | Price | Quantity & Availability | ||||
19-172-012
|
Cardiac Science
9300P1001P |
Defibrillator, IntelliSense battery (9146), pr of defibrillation pads, Quick Start Tool Kit | 3.2kg |
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. Part Not Found |
N/A |
Description
Designed for medically trained EMS responders. Uses a proprietary analysis algorithm RHYTHMx to proactively monitor for life-threatening arrhythmias but allows user to take control when necessary. Semi-automatic operation with manual override and three-lead ECG monitoring capability.
- Intuitive and comprehensive voice and text prompts guide the user through the rescue
- Full-color ECG display
- Infrared data communications
- Configurable biphasic energy protocols and detection rates
- STAR biphasic technology customizes defibrillation therapy for each patient
- Features include noncommitted shock, pacemaker detection, and synchronized shock
- Multiple-rescue internal memory storage
- Built-in metronome (which can be switched off) can set the pace for CPR compressions
- Daily, weekly and monthly self-tests of battery, internal electronics, software and pads
- Weekly, the AED partially charges the high-voltage electronics
- Monthly, the AED charges the high-voltage electronics to full energy
Warranty and Services
Seven years
Specifications
Automated External Defibrillator |
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.