Learn More
Advanced ImmunoChemical, Inc.Supplier Diversity Partner Cardiac Troponin I Calibrator Set  DFS Item
Manufacturer:  Advanced ImmunoChemical, Inc.Supplier Diversity Partner 8TISET
This item is manufactured by HyTest, Ltd and distributed by Advanced ImmunoChemical, Inc. Cardiac troponin I calibrator set. Cardiac troponin I calibrator set is intended for use as a reference & control material in clinical laboratory immunoassay of human cardiac troponin I.Calibrators correspond to 0, 0.5, 1, 3, 10, 50, 100 ng/ml cardiac troponin I. Exact concentrations are indicated on each vial. Other range of concentrations can be prepared on the request. Presented freeze-dried from solution of human native troponin complex (except for standard 0 ng/ml) in normal human serum.Human native troponin complex purified from human cardiac muscle: blood sample from the tissue donor was tested & found negative for HBsAg, HCV and HIV 1 + 2 antibodies.Human serum was tested & found negative for HBsAg, HCV and HIV 1 + 2 antibodies.Storage Conditions:Store unopened vials at -70 °C (between -65 and -80 °C allowed).
Catalog No. 50-153-9830