Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Cardiac Troponin C Polyclonal Antibody

Rabbit Polyclonal Antibody

Supplier:  Invitrogen™ PA551325

Catalog No. PA551325


Only null left
Add to Cart

Description

Description

The antibody detects endogenous levels of total Cardiac Troponin C protein.

Troponin (Tn) is the central regulatory protein of striated muscle contraction. Tn consists of three components: Tn-I which is the inhibitor of actomyosin ATPase, Tn-T which contains the binding site for tropomyosin and Tn-C. The binding of calcium to Tn-C abolishes the inhibitory action of Tn on actin filaments, and as a result of calcium-ion uptake by troponin-C, the elements of the muscle thin filament undergo a series of allosteric changes which allow the interaction of actin with myosin, the hydrolysis of ATP, and the generation of tension.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

Cardiac Troponin C
Polyclonal
Unconjugated
Tnnc1
AI874626; cardiac 3; cardiac troponin C; Cardiac troponin I; CMD1FF; CMD1Z; CMD2A; CMH13; CMH7; cTnC; cTnI; MGC116817; RCM1; slow twitch skeletal/cardiac muscle troponin C; Tn1; TnC; TN-C; Tncc; Tni; TNN I3; TNNC; TNNC 1; Tnnc1; TNNI3; troponin C type 1 (slow); troponin C, cardiac/slow skeletal; troponin C, slow skeletal and cardiac muscles; troponin C1, slow; troponin C1, slow skeletal and cardiac type; Troponin I; Troponin I cardiac; TroponinI
Rabbit
Antigen Affinity Chromatography
RUO
21924, 7134
-20°C
Liquid
Immunohistochemistry (Paraffin), Western Blot
1 mg/mL
PBS with 40% glycerol and 0.05% sodium azide; pH 7.4
P19123, P63316
Tnnc1
Full length fusion protein of human TNNC1.
100 μL
Primary
Human, Mouse
Antibody
IgG
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.