Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ Cardiac Troponin C Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA580171
Description
Reconstitute with 0.2 mL of distilled water to yield a concentration of 500 μg/mL. Positive Control - WB: Rat Skeletal Muscle Tissue. IHC: Rat Cardiac Muscle Tissue.
Troponin (Tn) is the central regulatory protein of striated muscle contraction. Tn consists of three components: Tn-I which is the inhibitor of actomyosin ATPase, Tn-T which contains the binding site for tropomyosin and Tn-C. The binding of calcium to Tn-C abolishes the inhibitory action of Tn on actin filaments, and as a result of calcium-ion uptake by troponin-C, the elements of the muscle thin filament undergo a series of allosteric changes which allow the interaction of actin with myosin, the hydrolysis of ATP, and the generation of tension.
Specifications
Cardiac Troponin C | |
Polyclonal | |
Unconjugated | |
Tnnc1 | |
AI874626; cardiac 3; cardiac troponin C; Cardiac troponin I; CMD1FF; CMD1Z; CMD2A; CMH13; CMH7; cTnC; cTnI; MGC116817; RCM1; slow twitch skeletal/cardiac muscle troponin C; Tn1; TnC; TN-C; Tncc; Tni; TNN I3; TNNC; TNNC 1; Tnnc1; TNNI3; troponin C type 1 (slow); troponin C, cardiac/slow skeletal; troponin C, slow skeletal and cardiac muscles; troponin C1, slow; troponin C1, slow skeletal and cardiac type; Troponin I; Troponin I cardiac; TroponinI | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
21924, 290561, 7134 | |
-20°C | |
Lyophilized |
Immunohistochemistry (Paraffin), Western Blot | |
500 μg/mL | |
PBS with 5mg BSA and 0.05mg sodium azide, 0.05mg thimerosal | |
P19123, P63316 | |
Tnnc1 | |
A synthetic peptide corresponding to a sequence at the N-terminus of human cardiac Troponin C (1-18aa MDDIYKAAVEQLTEEQKN). | |
100 μg | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction