Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Diazyme Laboratories Cardiac Troponin I Control

Diazyme Cardiac Tropoinin I Control set is for the testing of Cardiac Troponin I in human serum.

Manufacturer:  Diazyme Laboratories DZ145ACON

Catalog No. 23-700-547


Due to product restrictions, please to purchase or view availability for this product.

This item is not returnable. View return policy

Description

Description

  • Highly sensitive liquid stable latex enhanced immunoturbidimetric test kit, designed to work on most open clinical chemistry analyzers
  • Provides rapid reliable results at a low reagent cost
  • Convenient, fast, and an accurate alternative to more expensive methods that require a dedicated immunochemistry instrument
  • The assay offers reliable results across the entirely clinical important reportable range from 0.4 - 10.0 ng/mL troponin I with excellent precision CV% of <2.8% at 2.85 mg/mL
Specifications

Specifications

2 x 1 mL vial
Cardiac Troponin I
Consists of 2 vials
Store at 2 to 8°C. DO NOT FREEZE. Note: Store CONTROL tightly capped when not in use.
TROPONIN I ASSAY, CONTROL SET (liquid stable)
Stability is up to the expiration date on label when stored at 2 to 8°C.
USA: for Research Only, CE
SDS
Documents
Product Certifications
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit