Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Cardiac Troponin I Polyclonal Antibody

Goat Polyclonal Antibody

Supplier:  Invitrogen™ PA186820

Catalog No. PIPA186820


Only null left
Add to Cart

Description

Description

Goat anti Human Troponin I antibody recognizes human cardiac-specific troponin I, (TnI) an integral inhibitory protein of cardiac muscle which exists as a complex with troponin C (TnC) and troponin T (TnT).

Cardiac Troponin I (Cardiac-specific troponin I, TnI, Troponin I) is an integral inhibitory protein in cardiac muscle that exists as part of a complex with troponin C (TnC) and troponin T (TnT). The TnT subunit of troponin binds to tropomyosin-t form, a troponin-tropomyosin complex, anchored in place by the binding of TnI to actin, within muscle thin filaments. Structural change resulting from the binding of calcium to specific sites on the regulatory TnC subunit, releases the inhibitory region of TnI from actin, anabling the attachment of the molecular motor protein myosin, allowing for muscle contraction and hence movement. The measurements of blood TnI and TnT levels is an important diagnostics indicator of heart muscle damage, and can be used to differentiate between angina and myocardial infarction in patients with chest pains. Cardiac Troponin I has also been shown to inhibit angiogenesis in vivo and in vitro. Mutations in the Cardiac Troponin I gene cause familial hypertrophic cardiomyopathy type 7 (CMH7) and familial restrictive cardiomyopathy (RCM).
TRUSTED_SUSTAINABILITY
Specifications

Specifications

Cardiac Troponin I
Polyclonal
Unconjugated
TNNI3
alternatively spliced; cardiac ankyrin repeat kinase; Cardiac muscle troponin T; cardiac troponin I; cardiac troponin I isoform; cardiac troponin I-interacting kinase; cardiac troponin T; cardiac troponin T isoform 1; cardiac troponin T isoform 2; cardiac troponin T2; cardiomyopathy, dilated 1D (autosomal dominant); cardiomyopathy, dilated 2A (autosomal recessive); cardiomyopathy, hypertrophic 2; CARK; CCDD; CMD1D; CMD1FF; CMD2A; CMH2; CMH7; CMPD2; cTnI; cTnT; Ctt; CTTG; D830019J24Rik; LVNC6; MGC116817; RATCTTG; RCM1; RCM3; Serine/threonine-protein kinase TNNI3K; Tn1; TnI; TNNC1; Tnni3; TNNI3 interacting kinase; TNNI3-interacting kinase; TNNI3K; tnni3k {ECO:0000250; TNNT2; Tnnt3; TnTc; Tpn1; troponin 1, type 3; troponin I type 3 (cardiac); troponin I, cardiac 3; troponin I, cardiac muscle; troponin I, cardiac muscle-like; troponin I3, cardiac type; Troponin T cardiac; troponin T cardiac isoform; troponin T isoform 1; troponin T isoform 2; troponin T splicing mutant isoform 1; troponin T splicing mutant isoform 2; troponin T type 2 (cardiac); troponin T, cardiac muscle; troponin T2, cardiac; troponin T2, cardiac type; truncated cardiac troponin T; UniProtKB:Q59H18}
Goat
Affinity chromatography
RUO
7137
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Liquid
ELISA, Immunohistochemistry (Frozen), Western Blot, Immunocytochemistry
1 mg/mL
PBS with <0.1% sodium azide; pH 7.4
P19429
TNNI3
Native human cardiac troponin I, purity greater than 98%.
200 μg
Primary
Human
Antibody
IgG
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

WARNING: Cancer - www.P65Warnings.ca.gov
SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.