Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Cardiac Troponin T Mouse anti-Human, Clone: BGN/1288/44, Novus Biologicals™
Mouse Monoclonal Antibody
Supplier: Novus Biologicals NB10066490
Description
Troponin T Type 2 (cardiac) Monoclonal specifically detects Troponin T Type 2 (cardiac) in Human samples. It is validated for Western Blot, ELISA.Specifications
Cardiac Troponin T | |
Monoclonal | |
1.0 mg/mL | |
Western Blot 1:250, ELISA 1:100-1:2000 | |
Cardiac Muscle Troponin T, Cardiomyopathy, Dilated 1D (Autosomal Dominant), Cardiomyopathy, Hypertrophic, CMD1D, CMH2, CMPD2, cTnT, LVNC6, RCM3, TnTc, troponin T type 2 (cardiac), troponin T, cardiac muscle, Troponin T2, Cardiac Type, Truncated Cardiac Troponin T | |
Mouse | |
Protein A purified | |
RUO | |
7139 | |
Human | |
Purified |
Western Blot, ELISA | |
BGN/1288/44 | |
Unconjugated | |
P45379 | |
TNNT2 | |
HMW conjugate | |
0.1 mg | |
Primary | |
NB100-66490 recognises Troponin T Type 2 (cardiac) isoform 1. In Western blotting, this reacts with reduced purified cardiac isoform, but does not react with the skeletal isoform. | |
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. | |
IgG1 |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
For Research Use Only