Cardinal™ Esteem™ Blue Sterile Powder-Free Synthetic Polyisoprene Surgical Gloves

With Neu-Thera™ emollient coating which applied to the interior of the gloves to aid in skin wellness and hand hygiene

Manufacturer: CARDINAL HEALTH CORP 2D73EB80

 View more versions of this product

Catalog No. 19-262-113

  Check Availability
Add to Cart

    Specifications

    Certifications ASTM F 1671
    Glove Cuff Style Beaded interlocking
    For Use With (Application) healthcare professionals
    Length (English) 12 in.
    Latex Free Yes
    Length (Metric) 30.5cm
    Material Polyisoprene
    Powdered No
    Size 8
    Sterile Yes
    Thickness 7.9mil
    Coating Material Smooth
    Color Blue

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit