Cardinal Health Astound ™ Fabric Reinforced Gowns

Protection, comfort and convenience

Manufacturer: Dupont Personal Protection 9511

 View more versions of this product

Catalog No. 19-130-5568

 N/A
N/A

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. 


    Specifications

    Certifications/Compliance AAMI
    Color Blue
    For Use With (Application) Protection for the wearer from particles, fluids and light chemical splash
    Size Large
    Includes One hand towel per gown
    Sterile Yes
    Material Non-woven, Breathable Astound™

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit