Learn More
Cardinal Health™ Bio-Shield™ Wrap
Excellent barrier against airborne bacteria
Supplier:  Cardinal Health™ 4024
 View more versions of this product
Catalog No. 19-150-0012
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. View substitute products »
Description
Description
Excellent barrier against airborne bacteria
  • Soft, drapable wrap
  • Repels fluids
  • Virtually lint free
  • Protects wrapped contents from contamination
  • Regular Duty (1.0 oz../sq. yd.)
Specifications
Specifications
Blue
Wet Laid Cellulose
Non-sterile
24 x 24 in. (60.96 x 60.96 cm)
60.96 cm
60.96 cm
Excellent barrier against airborne bacteria, protects wrapped contents from contamination, ideal for lighter specialty trays, pitchers, small basins and huck towels
Bio-shield™ Regular Duty Wrap
500/Cs.
24 in.
24 in.