Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Cardinal Health™ Bio-Shield™ Wrap
Excellent barrier against airborne bacteria
Supplier: Cardinal Health™ 4054
Description
Excellent barrier against airborne bacteria
- Soft, drapable wrap
- Repels fluids
- Virtually lint free
- Protects wrapped contents from contamination
- Regular Duty (1.0 oz../sq. yd.)
Specifications
Blue | |
Wet Laid Cellulose | |
Non-sterile | |
54 x 54 in. (137.16 x 137.16 cm) | |
137.16 cm | |
137.16 cm |
Excellent barrier against airborne bacteria, protects wrapped contents from contamination, ideal for lighter specialty trays, pitchers, small basins and huck towels | |
Bio-shield™ Regular Duty Wrap | |
100/Cs. | |
54 in. | |
54 in. |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title