Cardinal Health CleanCare Soaps


 View all options for this product

Catalog No. 19-162-206


This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. 


    Size (English) 50.7 oz.
    Size (Metric) 1.49L
    Packaging Bottle
    Quantity Each
    Certifications/Compliance CDC hand hygiene guidelines
    Composition Triclosan 0.3%, no Petroleum based substances
    Product Type Amino+Derm™ Antibacterial Soap
    For Use With (Application) Effective against range of micoorganisms

  • Description & Specifications

Related Products

Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit