Learn More
Our pricing system is unavailable. You are viewing list price.
Please call 1-800-766-7000 to place your order or try our site again later.
Cardinal Health CR100™ Sterile Latex Powder-Free Cleanroom Gloves
Backed by the expertise of Cardinal Health, CR100™ Sterile Latex Powder-Free Cleanroom Gloves meet all relevant ASTM standards. Made from powder-free natural rubber latex.
$239.00
Specifications
Certifications/Compliance ANSI/AAMI/ISO13485, ASTM D 3577, ASTM D 5712
Color Natural
Length (English) 12 in.
Length (Metric) 30.5 cm
Material Rubber
View More Specs
Alert:
These products contain natural rubber latex, a component of which may cause allergic reaction in some users. Use only as directed and in accordance with manufacturer instructions and warnings. Please consult your institution's policies regarding use of these products.
Products 7
Catalog Number Mfr. No. Size Price Quantity & Availability  
Catalog Number Mfr. No. Size Price Quantity & Availability  
19-120-2995
View Documents
Cardinal Health
2Y1627
7.5
Case of 100 for $239.00
N/A
19-120-2994
View Documents
Cardinal Health
2Y1626
7
Case of 100 for $239.00
N/A
19-120-2993
View Documents
Cardinal Health
2Y1625
6.5
Case of 100 for $239.00
N/A
19-120-2996
View Documents
Cardinal Health
2Y1628
8
Case of 100 for $239.00
N/A
19-120-2992
View Documents
Cardinal Health
2Y1624
6
Case of 100 for $239.00
N/A
19-120-2997
View Documents
Cardinal Health
2Y1629
8.5
Case of 100 for $239.00
N/A
19-120-2998
View Documents
Cardinal Health
2Y1630
9
Case of 100 for $239.00
N/A
Description
Description
Recommended for ISO 4 / Class 10 or higher, Length 12.0IN; Nominal finger thickness 8.0MIL, Clean-processed and chlorinated, Sterile; powder-free; made with natural rubber latex, Exclusive hand molds with independent thumb design allow for natural fit and movement that helps reduce hand fatigue
Order Info
Performance tested and traceable with batch documentation on each pack. Certificate of Sterility available upon request.
Certifications
ASTM D 5712, ISO 13485
Compliance
Meet or exceed ASTM™ D 3577 physical property requirements.
Documentation is available upon request, Tested in accordance with ASTM D 5712, Contains 50 micrograms (μg) or less of total water extractable protein per square decimeter (/dm2), Caution: Safe use of these gloves by or on latex-sensitized individuals has not been established. This product contains natural rubber latex which may cause allergic reactions.
Specifications
Specifications
ANSI/AAMI/ISO13485, ASTM D 3577, ASTM D 5712
12 in.
Rubber
Yes
Powder-free
Natural
30.5 cm
Sterile
Fitted
7.5 mil