Learn More
Cardinal Health™ Gas Sterilization Pouches
Organize and package sterilized lab materials
Manufacturer:  CARDINAL HEALTH CORP T70714
 View more versions of this product
Catalog No. 19-160-180
  Check Availability
Add to cart
Specifications Description & Specifications
Specifications
35.6cm
17.8cm
For non-sterile, clean items, can be sterilized with end open for venting and sealed after processing, constructed with strong 2mil plastic film
14 in.
7 in.
Plastic
14 x 7 in. (35.6 x 17.8cm)
Green film
2 mil