Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Cardinal Health™ General-Purpose Coveralls With Tyvek™
Click to view available options
Size:
Large
X-Large
2X-Large
3X-Large
Description
- Feature full-length front zipper and elastic wrists and ankles
- Basic style offers set-in sleeves and traditional collar
- Premier style offers raglan sleeves, bound collar, and a more generous cut
- Both styles are white
Order Info
To ensure cleanliness, coveralls are packed in double poly-lined bags.
Specifications
Product Type | Cleanroom Coveralls |
Size | X-Large |
Material | Tyvek™ |
Color | White |
Disposable | Single-use |
Sterility | Non-sterile |
For Use With (Application) | Controlled Environments |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction