Learn More
Cardinal Health General-Purpose Coveralls With Tyvek
Available in two styles
Specifications
Color | White |
---|
Alert:
Garments made of Tyvek should not be used around heat, flames, sparks, or in potentially flammable or explosive environments.
Catalog Number | Mfr. No. | Size | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Size | Price | Quantity & Availability | |||||
19-130-2117
|
Cardinal Health
93793 |
3X-Large |
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. |
View substitute products » |
19-147-757
|
Cardinal Health
9377 |
Large |
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. |
View substitute products » |
19-147-758
|
Cardinal Health
9378 |
X-Large |
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. |
View substitute products » |
19-147-759
|
Cardinal Health
9379 |
2X-Large |
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. |
View substitute products » |
Description
- Feature full-length front zipper and elastic wrists and ankles
- Basic style offers set-in sleeves and traditional collar
- Premier style offers raglan sleeves, bound collar, and a more generous cut
- Both styles are white
Order Info
To ensure cleanliness, coveralls are packed in double poly-lined bags.
Specifications
White |