Learn More
Cardinal Health™ High-Top Shoe Covers with Tyvek™
Offer both durability and strength
Manufacturer:  Cardinal Health™ 9316
 View more versions of this product
Catalog No. 19-047-236
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
View substitute products »
Alert:
Garments made of Tyvek™ should not be used around heat, flames, sparks or in potentially flammable or explosive environments.
Description
Description
  • Made of low-linting Tyvek material
  • With skid-resistant vinyl soles
  • Adjustable ankle ties and latex-free elastic bands keep covers securely in place
  • Color: white
Order Info
Bulk-packaged in double polybags for added cleanliness.
Specifications
Specifications
Safety
White
Large
Tyvek™