Cardinal Health™ Low-Temperature Sterilization Pouchess
Protect the sterile integrity of tools and equipment
Manufacturer: CARDINAL HEALTH CORP 37812
 View more versions of this product
Catalog No. 19-160-125
  Check Availability
Add to Cart
    Specifications
    Material Tyvek™
    Dimensions (L x W) 20.3 × 30.5cm (8 × 12 in.)
    For Use With (Application) For low temperature sterilization, ethylene oxide and plasma
    Seal Type Heat seal
    Closure Type Heat-Seal
  • Description & Specifications
Related Products
Product Content Feedback