Cardinal Health™ Low-Temperature Sterilization Pouchess
Protect the sterile integrity of tools and equipment
Manufacturer: CARDINAL HEALTH CORP 32218
 View more versions of this product
Catalog No. 19-160-111
  Check Availability
Add to Cart
    Specifications
    Closure Type Self seal
    Dimensions (L x W) 30.5 × 45.7cm (12 × 18 in.)
    For Use With (Application) Excellent choice for devices requiring low temperature sterilization, produced without an indicator for all types of low temperature sterilization, including EO and plasma
    Material Tyvek™
  • Description & Specifications
Related Products
Product Content Feedback