Learn More
Cardinal Health™ Medi-Vac™ Guardian™ Suction Canisters
With automatic overflow shutoff valve located inside lid to help prevent cross contamination of regulators and wall vacuum outlets
Manufacturer:  CARDINAL HEALTH CORP 65651220
 View more versions of this product
Catalog No. 19-162-320
  Check Availability
Add to cart
Specifications Description & Specifications
Specifications
2L
Overflow shutoff valve, 90° adapters, accessory and orthopedic port
1.25 lb.
Suction canister
0.56kg
67.63 oz., 2L