Learn More
Cardinal Health™ Snap-up Frock with Tyvek™
Full-length frock snaps all the way to the neck for maximum protection
Manufacturer: Cardinal Health™ 9339
Alert:
Garments made of Tyvek™ should not be used around heat, flames, sparks, or in potentially flammable or explosive environments.
Description
- Features adjustable high tab collar
- No pockets to trap dirt or lint
- Basic style offers standard fit with set-in sleeves
- Premier style offers raglan sleeves with a more generous cut
- Packed in double poly liners to ensure cleanliness
Specifications
2X-Large | |
Tyvek™ | |
White |