Learn More
Cardinal Health Snap-up Frock with Tyvek
Full-length frock snaps all the way to the neck for maximum protection
Specifications
Color | White |
---|---|
Material | Tyvek™ |
Alert:
Garments made of Tyvek should not be used around heat, flames, sparks, or in potentially flammable or explosive environments.
Catalog Number | Mfr. No. | Description | Price | Qty | Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Description | Price | Qty | Availability | ||||
19-130-2013
|
Cardinal Health
9339 |
N/A | Set-in Sleeves; XX-Large |
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. |
View substitute products » |
19-130-2124
|
Cardinal Health
9347 |
N/A | Set-in Sleeves; 4X-Large |
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. |
N/A |
Description
- Features adjustable high tab collar
- No pockets to trap dirt or lint
- Basic style offers standard fit with set-in sleeves
- Premier style offers raglan sleeves with a more generous cut
- Packed in double poly liners to ensure cleanliness
Specifications
White | |
Tyvek™ |