Cardinal Health™ Sterile CP100™ BT Latex Clean Process Gloves
Extremely comfortable “flex and stretch” design
Manufacturer: CARDINAL HEALTH CORP 2Y5006
 View more versions of this product
Catalog No. 19-120-2973
  Check Availability
Add to Cart
Alert
Warning: These products contain natural rubber latex, a component of which may cause allergic reaction in some users. Use only as directed and in accordance with manufacturer instructions and warnings. Please consult your institution's policies regarding use of these products.
    Specifications
    Certifications ANSI/AAMI/ISO, ISO13485, ASTM D 5712
    Glove Cuff Style Beaded
    Length (English) 12 in.
    Length (Metric) 30.5cm
    Material Natural Rubber
    Powdered No
    Size 8.5
    Sterile Yes
    Thickness 7mil
    Disposable Yes
    Hand Hand-specific
    Coating Material Anti-slip textured
    Color Natural
  • Description & Specifications
Related Products
Product Content Feedback