Learn More
Cardinal Health CR100 Sterile Nitrile Powder-Free Cleanroom Gloves
Backed by the expertise of Cardinal Health, CR100 Sterile Nitrile Powder-Free Cleanroom Gloves meet all relevant ASTM standards. Made from powder-free, synthetic nitrile.
Catalog Number | Mfr. No. | Certifications/Compliance | Size | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Certifications/Compliance | Size | Price | Quantity & Availability | ||||
19-130-5619
|
Cardinal Health
2Y1860T |
ASTM D 6319, ISO Class 4 | 5.5 |
Case of 100 for $202.00
|
N/A | ||||
Due to product restrictions, please Sign In to purchase or view availability for this product. |
|||||||||
19-130-5620
|
Cardinal Health
2Y1861T |
ASTM D 6319 | 6 |
Case of 100 for $202.00
|
N/A | ||||
Due to product restrictions, please Sign In to purchase or view availability for this product. |
|||||||||
19-130-5626
|
Cardinal Health
2Y1867T |
ASTM D 6319 | 9 |
Case of 100 for $202.00
|
N/A | ||||
Due to product restrictions, please Sign In to purchase or view availability for this product. |
|||||||||
19-130-5622
|
Cardinal Health
2Y1863T |
ASTM D 6319 | 7 |
Case of 100 for $202.00
|
N/A | ||||
Due to product restrictions, please Sign In to purchase or view availability for this product. |
|||||||||
19-130-5623
|
Cardinal Health
2Y1864T |
ASTM D 6319 | 7.5 |
Case of 100 for $202.00
|
N/A | ||||
Due to product restrictions, please Sign In to purchase or view availability for this product. |
|||||||||
19-130-5625
|
Cardinal Health
2Y1866T |
ASTM D 6319 | 8.5 |
Case of 100 for $202.00
|
N/A | ||||
Due to product restrictions, please Sign In to purchase or view availability for this product. |
|||||||||
19-130-5621
|
Cardinal Health
2Y1862T |
ASTM D 6319 | 6.5 |
Case of 100 for $202.00
|
N/A | ||||
Due to product restrictions, please Sign In to purchase or view availability for this product. |
|||||||||
19-130-5624
|
Cardinal Health
2Y1865T |
ASTM D 6319 | 8 |
Case of 100 for $202.00
|
N/A | ||||
Due to product restrictions, please Sign In to purchase or view availability for this product. |
|||||||||
Description
Recommended for ISO 4 / Class 10 or higher, Length 11.6IN; Nominal finger thickness 3.9MIL, Clean-processed and chlorinated, Beaded cuff design helps reduce roll-down; micro-textured for confident and consistent grip, Packed in an ISO 5 / Class 100 cleanroom environmentOrder Info
Clean-processed and packaged in an ISO Class 4 (Class 10) environment. Pair packed in a polyfilm inner wallet and heat-sealed into an outer polybag.
Certifications
ISO 13485
Compliance
Meet or exceed ASTM D 6319 requirements.
Documentation is available upon request
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.