Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cardinal Health™ Ultrafree™ Max Powder-Free Latex Surgical Gloves

Offer extra protection without sacrificing comfort

Manufacturer:  CARDINAL HEALTH CORP 2D7292I

 View more versions of this product

Catalog No. 19-130-5877


  Check Availability
Add to cart

Specifications Description & Specifications

Specifications

Smooth, anti-slip
Beaded/Rolled
Rubber, natural
11.3mil
Powder-free
Sterile
30.5cm
Yes
Hand-specific
6.5
ASTM D 5712
Dark Brown
12 in.
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit