Learn More
3B Scientific™ Cardionics PAT Basic™ Pediatric Auscultation Trainer Products for Science Education
3B Scientific™ Cardionics PAT Basic™ Pediatric Auscultation Trainer with WiFi engages SimScope™ stethoscope technology to simulate pediatric sounds
$7000.00
Specifications
For Use With (Application) Pediatric-Specific Disorders
Includes SimScope™ Stethoscope Technology
Product Type Manikin
Weight (English) 9.91 lb.
View More Specs
Products 1
Catalog Number Mfr. No. Color Price Quantity & Availability  
Catalog Number Mfr. No. Color Price Quantity & Availability  
S27951 Products for Science Education
View Documents
3B Scientific™
1022472
Dark Skin Each for $7,000.00
Add to cart
Description
Description
  • The PAT Basic™ trainer engages the technology of the SimScope™ stethoscope to simulate pediatric heart, lung, and bowel sounds utilizing a large sounds library
  • Using the provided SimScope stethoscope, users are able to access a large variety of pathological conditions in order to customize scenarios
  • By placing the SimScope over the anatomically correct locations of PAT Basic, the programmed sounds will be played through the scope, allowing for students to employ and enhance their diagnostic skills
  • The SimScope WiFi upgrade enables wireless communication between the Simscope and PAT BasicC manikin which allows users to change sounds remotely or to use in conjunction with a scenario-based simulation
  • PAT Basic consists of a Male Manikin, SimScope WiFi, SimScope Software, Site License, USB cable, SAM T-Shirt, and a User's Manual
Specifications
Specifications
Pediatric-Specific Disorders
SimScope™ Stethoscope Technology
Manikin
9.91 lb.